Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204009.1 | NP_001190938.1 | 5 - 6 | 706 | 94 | |
NM_119856.3 | NP_195410.1 | 4 - 5 | 865 | 94 | ||
GenBank mRNA | U12546.1 | - | 4 - 5 | 780 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |