Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204009.1 | NP_001190938.1 | 10 - 11 | 1181 | 69 | |
NM_119856.3 | NP_195410.1 | 9 - 10 | 1340 | 69 | ||
GenBank mRNA | U12546.1 | - | 9 - 10 | 1255 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |