Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119857.3 | NP_568010.1 | 1 - 2 | 593 | 81 | |
GenBank mRNA | AF319540.1 | - | 1 - 2 | 531 | 81 | |
AK229267.1 | - | 1 - 2 | 549 | 81 | ||
BT026462.1 | - | 1 - 2 | 509 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |