Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119857.3 | NP_568010.1 | 3 - 4 | 773 | 150 | |
GenBank mRNA | AF319540.1 | - | 3 - 4 | 711 | 150 | |
AK229267.1 | - | 3 - 4 | 729 | 150 | ||
BT026462.1 | - | 3 - 4 | 689 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |