Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119857.3 | NP_568010.1 | 4 - 5 | 834 | 89 | |
GenBank mRNA | AF319540.1 | - | 4 - 5 | 772 | 89 | |
AK229267.1 | - | 4 - 5 | 790 | 89 | ||
BT026462.1 | - | 4 - 5 | 750 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |