Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAPRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119858.6 | NP_195412.3 | 3 - 4 | 498 | 124 | |
GenBank mRNA | AY093004.1 | - | 3 - 4 | 466 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |