Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119867.3 | NP_568014.1 | 3 - 4 | 485 | 61 | |
GenBank mRNA | AF250963.1 | - | 3 - 4 | 446 | 61 | |
AK118314.1 | - | 3 - 4 | 437 | 61 | ||
AY085839.1 | - | 3 - 4 | 443 | 61 | ||
BT005680.1 | - | 3 - 4 | 410 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |