Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119867.3 | NP_568014.1 | 5 - 6 | 685 | 142 | |
GenBank mRNA | AF250963.1 | - | 5 - 6 | 646 | 142 | |
AK118314.1 | - | 5 - 6 | 637 | 142 | ||
AY085839.1 | - | 5 - 6 | 643 | 142 | ||
BT005680.1 | - | 5 - 6 | 610 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |