Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119890.7 | NP_195444.1 | 9 - 10 | 1997 | 66 | |
GenBank mRNA | AJ872069.1 | - | 9 - 10 | 1829 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |