Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP81D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119898.4 | NP_195451.1 | 2 - 3 | 1017 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |