Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP91A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119906.3 | NP_195459.1 | 1 - 2 | 1014 | 85 | |
GenBank mRNA | AY039844.1 | - | 1 - 2 | 941 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |