Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYCD5;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342450.1 | NP_001320153.1 | 3 - 4 | 772 | 117 | |
NM_001342451.1 | NP_001328589.1 | 3 - 4 | 963 | 117 | ||
NM_119926.5 | NP_195478.2 | 3 - 4 | 963 | 117 | ||
GenBank mRNA | AK221431.1 | - | 3 - 4 | 852 | 117 | |
BT011732.1 | - | 3 - 4 | 634 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |