Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119948.4 | NP_195500.1 | 2 - 3 | 561 | 65 | |
GenBank mRNA | AF372922.1 | - | 2 - 3 | 390 | 65 | |
AK226329.1 | - | 2 - 3 | 397 | 65 | ||
AY078035.1 | - | 1 - 2 | 278 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |