Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mtHsc70-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342461.1 | NP_001328002.1 | 4 - 5 | 883 | 118 | |
NM_119952.4 | NP_195504.2 | 4 - 5 | 871 | 118 | ||
GenBank mRNA | BT002390.1 | - | 4 - 5 | 868 | 118 | |
BT008430.1 | - | 4 - 5 | 790 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |