Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAUT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119989.5 | NP_195540.2 | 5 - 6 | 389 | 93 | |
GenBank mRNA | KJ138714.1 | - | 5 - 6 | 328 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |