Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSL03 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202724.2 | NP_001189653.1 | 27 - 28 | 3740 | 93 | |
NM_001336361.1 | NP_001318331.1 | 28 - 29 | 3452 | 93 | ||
NM_001336362.1 | NP_001323730.1 | 28 - 29 | 3206 | 93 | ||
NM_001336363.1 | NP_001323731.1 | 28 - 29 | 3377 | 93 | ||
NM_179847.3 | NP_850178.2 | 27 - 28 | 3380 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |