Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSP60-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202736.1 | NP_001189665.1 | 4 - 5 | 528 | 92 | |
NM_179872.3 | NP_850203.1 | 4 - 5 | 556 | 92 | ||
GenBank mRNA | AK175415.1 | - | 4 - 5 | 515 | 92 | |
AY136360.1 | - | 4 - 5 | 392 | 92 | ||
BT000103.1 | - | 4 - 5 | 352 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |