Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSP60-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179872.3 | NP_850203.1 | 8 - 9 | 1113 | 83 | |
GenBank mRNA | AK175415.1 | - | 8 - 9 | 1072 | 83 | |
AY136360.1 | - | 8 - 9 | 949 | 83 | ||
BT000103.1 | - | 8 - 9 | 909 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |