Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036413.3 | NP_001031490.1 | 3 - 4 | 1766 | 62 | |
NM_129153.3 | NP_181138.1 | 4 - 5 | 1548 | 62 | ||
NM_179925.3 | NP_850256.1 | 3 - 4 | 1422 | 62 | ||
GenBank mRNA | AK227272.1 | - | 4 - 5 | 1485 | 62 | |
AY072175.1 | - | 4 - 5 | 1428 | 62 | ||
AY096548.1 | - | 2 - 3 | 1223 | 62 | ||
AY099854.1 | - | 3 - 4 | 1420 | 62 | ||
BT008422.1 | - | 2 - 3 | 1223 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |