Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179940.6 | NP_850271.5 | 30 - 31 | 3483 | 72 | |
GenBank mRNA | GQ373182.1 | - | 30 - 31 | 3185 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |