Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179940.6 | NP_850271.5 | 5 - 6 | 1001 | 86 | |
GenBank mRNA | GQ373182.1 | - | 5 - 6 | 703 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |