Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129373.4 | NP_181352.1 | 6 - 7 | 1082 | 114 | |
NM_179961.2 | NP_850292.2 | 6 - 7 | 1032 | 114 | ||
NM_201901.4 | NP_973630.1 | 6 - 7 | 1041 | 114 | ||
GenBank mRNA | AF372938.1 | - | 6 - 7 | 1029 | 114 | |
AF461691.1 | - | 6 - 7 | 969 | 114 | ||
AK316894.1 | - | 6 - 7 | 1033 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |