Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035850.3 | NP_001030927.1 | 1 - 2 | 342 | 65 | |
NM_001331269.1 | NP_001322867.1 | 2 - 3 | 255 | 65 | ||
NM_001331270.1 | NP_001322868.1 | 2 - 3 | 225 | 65 | ||
NM_100019.2 | NP_563627.1 | 2 - 3 | 255 | 65 | ||
GenBank mRNA | AB081500.1 | - | 1 - 2 | 67 | 65 | |
AF465800.1 | - | 1 - 2 | 58 | 65 | ||
AK175840.1 | - | 1 - 2 | 114 | 65 | ||
BT024771.1 | - | 1 - 2 | 58 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |