Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KEA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331307.1 | NP_001320631.1 | 5 - 6 | 2359 | 105 | |
NM_100062.5 | NP_171684.2 | 6 - 7 | 2383 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |