Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160828.1 | NP_001154300.1 | 17 - 18 | 2444 | 86 | |
NM_001160829.1 | NP_001154301.1 | 18 - 19 | 2558 | 86 | ||
NM_001331317.1 | NP_001320318.1 | 16 - 17 | 2637 | 86 | ||
NM_100075.8 | NP_171697.3 | 17 - 18 | 2558 | 86 | ||
GenBank mRNA | AB290929.1 | - | 17 - 18 | 2407 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |