Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIMIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100126.4 | NP_563653.2 | 1 | 460 | 126 | |
GenBank mRNA | AK119149.1 | - | 1 | 438 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |