Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSLD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100153.4 | NP_171773.1 | 2 - 3 | 1949 | 92 | |
GenBank mRNA | KJ139033.1 | - | 2 - 3 | 1765 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |