Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100158.3 | NP_171778.1 | 3 - 4 | 1114 | 152 | |
GenBank mRNA | AF428425.1 | - | 3 - 4 | 1084 | 152 | |
AY065210.1 | - | 3 - 4 | 1084 | 152 | ||
AY133812.1 | - | 3 - 4 | 1015 | 152 | ||
X72291.1 | - | 1072 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |