Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100173.4 | NP_171791.1 | 2 - 3 | 474 | 108 | |
GenBank mRNA | AY062642.1 | - | 2 - 3 | 277 | 108 | |
AY093281.1 | - | 2 - 3 | 199 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |