Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FZL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083983.1 | NP_001077452.1 | 6 - 7 | 1515 | 122 | |
NM_001331388.1 | NP_001322638.1 | 6 - 7 | 1536 | 122 | ||
NM_100198.5 | NP_171815.3 | 6 - 7 | 1536 | 122 | ||
GenBank mRNA | AK176297.1 | - | 6 - 7 | 1508 | 122 | |
AK227660.1 | - | 6 - 7 | 1508 | 122 | ||
DQ462573.1 | - | 6 - 7 | 1515 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |