Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG18G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100220.8 | NP_171837.7 | 6 - 7 | 2694 | 65 | |
GenBank mRNA | BT002410.1 | - | 6 - 7 | 2635 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |