Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRP39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197976.1 | NP_001184905.1 | 2 - 3 | 452 | 63 | |
NM_001331460.1 | NP_001322270.1 | 1 - 2 | 507 | 63 | ||
NM_100289.4 | NP_563700.1 | 2 - 3 | 421 | 63 | ||
GenBank mRNA | AY056321.1 | - | 2 - 3 | 386 | 63 | |
AY087878.1 | - | 2 - 3 | 388 | 63 | ||
AY091187.1 | - | 1 - 2 | 238 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |