Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IAA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100291.3 | NP_171906.1 | 3 - 4 | 1056 | 91 | |
GenBank mRNA | AF332396.1 | - | 3 - 4 | 717 | 91 | |
U18412.1 | - | 3 - 4 | 1022 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |