Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331470.1 | NP_001322162.1 | 1 - 2 | 552 | 149 | |
NM_100305.4 | NP_171920.1 | 1 - 2 | 414 | 149 | ||
GenBank mRNA | AF332393.1 | - | 1 - 2 | 182 | 149 | |
AY064007.1 | - | 1 - 2 | 278 | 149 | ||
U18406.1 | - | 1 - 2 | 267 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |