Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331549.1 | NP_001320371.1 | 3 - 4 | 1107 | 91 | |
NM_100425.4 | NP_172037.1 | 3 - 4 | 1114 | 91 | ||
GenBank mRNA | AK117698.1 | - | 3 - 4 | 1107 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |