Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARI12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197990.1 | NP_001184919.1 | 12 - 13 | 1270 | 96 | |
NM_001331600.1 | NP_001322817.1 | 12 - 13 | 1270 | 96 | ||
NM_100469.2 | NP_172079.2 | 12 - 13 | 1249 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |