Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100512.3 | NP_172120.2 | 1 - 2 | 619 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |