Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CalS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331632.1 | NP_001322002.1 | 31 - 32 | 4119 | 100 | |
NM_100528.3 | NP_172136.2 | 31 - 32 | 3777 | 100 | ||
GenBank mRNA | HM049631.1 | - | 31 - 32 | 3735 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |