Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSBP-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100545.7 | NP_172153.1 | 1 - 2 | 556 | 87 | |
GenBank mRNA | AY056416.1 | - | 1 - 2 | 305 | 87 | |
AY070469.1 | - | 1 - 2 | 305 | 87 | ||
AY074308.1 | - | 1 - 2 | 307 | 87 | ||
AY087305.1 | - | 1 - 2 | 312 | 87 | ||
AY096487.1 | - | 1 - 2 | 250 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |