Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAUT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197996.1 | NP_001184925.1 | 4 - 5 | 684 | 131 | |
NM_100555.3 | NP_563771.1 | 4 - 5 | 600 | 131 | ||
GenBank mRNA | AY045900.1 | - | 4 - 5 | 532 | 131 | |
AY091452.1 | - | 4 - 5 | 267 | 131 | ||
KJ139026.1 | - | 4 - 5 | 267 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |