Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAUT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197996.1 | NP_001184925.1 | 5 - 6 | 762 | 124 | |
NM_100555.3 | NP_563771.1 | 5 - 6 | 678 | 124 | ||
GenBank mRNA | AY045900.1 | - | 5 - 6 | 610 | 124 | |
AY091452.1 | - | 5 - 6 | 345 | 124 | ||
KJ139026.1 | - | 5 - 6 | 345 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |