Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bZIP52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100562.3 | NP_172170.1 | 2 - 3 | 782 | 66 | |
GenBank mRNA | AJ419852.1 | - | 2 - 3 | 597 | 66 | |
AY062474.1 | - | 2 - 3 | 752 | 66 | ||
BT000102.1 | - | 2 - 3 | 597 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |