Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bZIP52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100562.3 | NP_172170.1 | 3 - 4 | 860 | 71 | |
GenBank mRNA | AJ419852.1 | - | 3 - 4 | 675 | 71 | |
AY062474.1 | - | 3 - 4 | 830 | 71 | ||
BT000102.1 | - | 3 - 4 | 675 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |