Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112954.4 | NP_566666.2 | 8 - 9 | 1294 | 84 | |
GenBank mRNA | AK226894.1 | - | 8 - 9 | 1144 | 84 | |
AX765820.1 | - | 8 - 9 | 1087 | 84 | ||
AY140096.1 | - | 8 - 9 | 1149 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |