Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112965.1 | NP_188710.1 | 12 - 13 | 1248 | 59 | |
GenBank mRNA | AF129516.1 | - | 12 - 13 | 1248 | 59 | |
AK117114.1 | - | 12 - 13 | 1105 | 59 | ||
BT005326.1 | - | 12 - 13 | 1050 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |