Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112965.1 | NP_188710.1 | 3 - 4 | 566 | 74 | |
GenBank mRNA | AF129516.1 | - | 3 - 4 | 566 | 74 | |
AK117114.1 | - | 3 - 4 | 423 | 74 | ||
BT005326.1 | - | 3 - 4 | 368 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |