Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGP40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112978.1 | NP_566668.1 | 1 | 241 | 70 | |
GenBank mRNA | AY086220.1 | - | 1 | 241 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |