Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFU4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035658.1 | NP_001030735.1 | 3 - 4 | 642 | 81 | |
NM_112989.3 | NP_566673.1 | 3 - 4 | 682 | 81 | ||
GenBank mRNA | AF370537.1 | - | 3 - 4 | 642 | 81 | |
AJ512936.1 | - | 3 - 4 | 588 | 81 | ||
AY072509.1 | - | 3 - 4 | 562 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |