Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHL-CPN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130029.4 | NP_566022.1 | 1 - 2 | 692 | 91 | |
GenBank mRNA | AB051163.1 | - | 1 - 2 | 145 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |