Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130069.3 | NP_182031.1 | 1 - 2 | 478 | 69 | |
GenBank mRNA | AB493590.1 | - | 1 - 2 | 217 | 69 | |
AK317134.1 | - | 1 - 2 | 292 | 69 | ||
BT000921.1 | - | 1 - 2 | 217 | 69 | ||
Y13649.1 | - | 255 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |